Skip to Content
Merck
All Photos(1)

Key Documents

250279

Sigma-Aldrich

3-(Trifluoromethyl)benzoyl chloride

98%

Synonym(s):

α,α,α-Trifluoro-m-toluoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4COCl
CAS Number:
Molecular Weight:
208.56
Beilstein:
391266
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.477 (lit.)

bp

184-186 °C/750 mmHg (lit.)

density

1.383 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

FC(F)(F)c1cccc(c1)C(Cl)=O

InChI

1S/C8H4ClF3O/c9-7(13)5-2-1-3-6(4-5)8(10,11)12/h1-4H

InChI key

RUJYJCANMOTJMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Trifluoromethyl)benzoyl chloride has been used in the preparation of intermediates, required for synthesis of C-2 and C-3 substituted pyrazolo[1,5-a]pyrimidines.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan M Berger et al.
Bioorganic & medicinal chemistry letters, 19(23), 6519-6523 (2009-10-30)
As part of our research effort to discover B-Raf kinase inhibitors, we prepared a series of C-3 substituted N-(3-(pyrazolo[1,5-a]pyrimidin-7-yl)phenyl)-3-(trifluoromethyl)benzamides. X-ray crystallography studies revealed that one of the more potent inhibitors (10n) bound to B-Raf kinase without forming a hinge-binding hydrogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service