Skip to Content
Merck
All Photos(1)

Key Documents

111902

Sigma-Aldrich

4-Chlorobenzoyl chloride

99%

Synonym(s):

4-Chlorobenzoic acid chloride, p-Chlorobenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4COCl
CAS Number:
Molecular Weight:
175.01
Beilstein:
471606
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050512
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.578 (lit.)

bp

102-104 °C/11 mmHg (lit.)

mp

11-14 °C (lit.)

density

1.365 g/mL at 20 °C (lit.)

functional group

acyl chloride
chloro

SMILES string

ClC(=O)c1ccc(Cl)cc1

InChI

1S/C7H4Cl2O/c8-6-3-1-5(2-4-6)7(9)10/h1-4H

InChI key

RKIDDEGICSMIJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorobenzoyl chloride is used as an intermediate compound in the synthesis of pharmaceuticals.

Application

4-Chlorobenzoyl chloride was used to synthesize 4-chlorobenzoyl CoA by reacting it with CoA in KHCO3 buffer.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Competing intramolecular NH? OC hydrogen bonds and extended intermolecular network in 1-(4-chlorobenzoyl)-3-(2-methyl-4-oxopentan-2-yl) thiourea analyzed by experimental and theoretical methods
Saeed, Aamer, et al.
Chemical Physics, 431, 39-46 (2014)
Iteb Trabelsi et al.
Journal of oleo science, 66(7), 667-676 (2017-07-05)
The article deals with the use of mixed anhydrides for the synthesis of fatty esters. Both aliphatic and aromatic acids are involved, indicating different behaviors according to the chain length of the aliphatic acid. We describe a novel and efficient
S D Copley et al.
Applied and environmental microbiology, 58(4), 1385-1387 (1992-04-01)
4-Chlorobenzoate degradation in cell extracts of Acinetobacter sp. strain 4-CB1 occurs by initial synthesis of 4-chlorobenzoyl coenzyme A (4-chlorobenzoyl CoA) from 4-chlorobenzoate, CoA, and ATP. 4-Chlorobenzoyl CoA is dehalogenated to 4-hydroxybenzoyl CoA. Following the dehalogenation reaction, 4-hydroxybenzoyl CoA is hydrolyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service