Merck
  • Role of Calbindin-D28k in Diabetes-Associated Advanced Glycation End-Products-Induced Renal Proximal Tubule Cell Injury.
Role of Calbindin-D28k in Diabetes-Associated Advanced Glycation End-Products-Induced Renal Proximal Tubule Cell Injury.
Cells (2019-07-03)
Kuo-How Huang, Siao-Syun Guan, Wei-Han Lin, Cheng-Tien Wu, Meei-Ling Sheu, Chih-Kang Chiang, Shing-Hwa Liu
ABSTRACT
Diabetes-associated advanced glycation end-products (AGEs) can increase extracellular matrix (ECM) expression and induce renal fibrosis. Calbindin-D28k, which plays a role in calcium reabsorption in renal distal convoluted tubules, is increased in a diabetic kidney. The role of calbindin-D28k in diabetic nephropathy still remains unclear. Here, calbindin-D28k protein expression was unexpectedly induced in the renal tubules of db/db diabetic mice. AGEs induced the calbindin-D28k expression in human renal proximal tubule cells (HK2), but not in mesangial cells. AGEs induced the expression of fibrotic molecules, ECM proteins, epithelial-mesenchymal transition (EMT) markers, and endoplasmic reticulum (ER) stress-related molecules in HK2 cells, which could be inhibited by a receptor for AGE (RAGE) neutralizing antibody. Calbindin-D28k knockdown by siRNA transfection reduced the cell viability and obviously enhanced the protein expressions of fibrotic factors, EMT markers, and ER stress-related molecules in AGEs-treated HK2 cells. Chemical chaperone 4-Phenylbutyric acid counteracted the AGEs-induced ER stress and ECM and EMT markers expressions. Calbindin-D28k siRNA in vivo delivery could enhance renal fibrosis in db/db diabetic mice. These findings suggest that inducible calbindin-D28k protects against AGEs/RAGE axis-induced ER stress-activated ECM induction and cell injury in renal proximal tubule cells.