Skip to Content
Merck
All Photos(1)

Key Documents

244953

Sigma-Aldrich

Potassium trichloro(ethylene)platinate(II) hydrate

Synonym(s):

Zeise’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K[(H2C=CH2)PtCl3] · xH2O
CAS Number:
Molecular Weight:
368.59 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

composition

Degree of hydration, ≤1

Quality Level

reaction suitability

core: platinum
reagent type: catalyst

mp

220 °C (dec.) (lit.)

density

2.88 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O.[K+].C=C.Cl[Pt-](Cl)Cl

InChI

1S/C2H4.3ClH.K.H2O.Pt/c1-2;;;;;;/h1-2H2;3*1H;;1H2;/q;;;;+1;;+2/p-3

InChI key

DCEGWIMEFFONKJ-UHFFFAOYSA-K

General description

Potassium trichloro(ethylene)platinate(II) hydrate is a transition-metal alkene complex commonly known as Zeise′s salt. It was first reported in the year 1930 and considered as a prototype for the classic Dewar-Chatt-Duncanson model.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of high external pressures on the vibrational spectra of Zeise?s complexes, K [Pt(η2-C2H4) Cl3] and [Pt(η2-C2H4) Cl2]2
Baldwin JA, et al.
Journal of Organometallic Chemistry, 690(13), 3165-3168 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service