Skip to Content
Merck
All Photos(1)

Key Documents

238112

Sigma-Aldrich

1,2-Diiodobenzene

98%

Synonym(s):

1,2-Phenylene diiodide, o-Diiodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4I2
CAS Number:
Molecular Weight:
329.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.718 (lit.)

bp

152 °C/15 mmHg (lit.)

density

2.524 g/mL at 25 °C (lit.)

functional group

iodo

storage temp.

2-8°C

SMILES string

Ic1ccccc1I

InChI

1S/C6H4I2/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

BBOLNFYSRZVALD-UHFFFAOYSA-N

General description

Photolysis of 1,2-diiodobenzene in cyclohexane, furan, benzene and benzene containing tetraphenylcyclopentadienone has been studied.

Application

1,2-Diiodobenzene has been used in the preparation of purino[8,9-f]phenanthridines.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor Cerna et al.
The Journal of organic chemistry, 75(7), 2302-2308 (2010-03-05)
Intramolecular C-H arylations were employed as a key step in the synthesis of hitherto unknown fused purine systems: 13-substituted purino[8,9-f]phenanthridines and 11-substituted 5,6-dihydropurino[8,9-a]isoquinolines. The purino[8,9-f]phenanthridines were prepared in moderate yields by double C-H arylations of 9-phenylpurines with 1,2-diiodobenzene or, more
The Photolysis of 1, 2-Diiodobenzene: A Photochemical Source of Benzyne.
Kampmeier JA and Hoffmeister E.
Journal of the American Chemical Society, 84(19), 3787-3788 (1962)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service