Skip to Content
Merck
All Photos(2)

Key Documents

233633

Sigma-Aldrich

4-Ethylbenzaldehyde

98%

Synonym(s):

p-Ethylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4CHO
CAS Number:
Molecular Weight:
134.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

221 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(CC)cc1

InChI

1S/C9H10O/c1-2-8-3-5-9(7-10)6-4-8/h3-7H,2H2,1H3

InChI key

QNGNSVIICDLXHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Ethylbenzaldehyde is a by-product of disinfection. Kinetic constants (KI) of 4-ethylbenzaldehyde for inhibition of the diphenolase activity of mushroom tyrosinase has been investigated.

Application

4-Ethylbenzaldehyde has been used in the synthesis of 4,4′-diaminotriphenylmethanes under microwave irradiation, which is useful for parallel library syntheses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted synthesis of 4, 4'-diaminotriphenylmethanes.
Guzman-Lucero D, et al.
Tetrahedron Letters, 46(7), 1119-1122 (2005)
M Jiménez et al.
Journal of agricultural and food chemistry, 49(8), 4060-4063 (2001-08-22)
A kinetic study of the inhibition of mushroom tyrosinase by 4-substituted benzaldehydes showed that these compounds behave as classical competitive inhibitors, inhibiting the oxidation of L-3,4-dihydroxyphenylalanine (L-DOPA) by mushroom tyrosinase (o-diphenolase activity). The kinetic parameter (K(I)) characterizing this inhibition was
Gergely Rácz et al.
Pathology oncology research : POR, 18(3), 579-584 (2011-12-14)
Disinfection of raw water is essential to the production of drinking water. However, by-products of disinfection may exert toxic effects. The potential toxic effects of two of these compounds, 4-ethylbenzaldehyde (EBA) and 2,4-difluoroaniline (DFA) were investigated using the zebrafish (Danio
Yunzi Feng et al.
Food chemistry, 265, 274-280 (2018-06-10)
Two types of chicken broth, broiler broth (BB) and native chicken broth (NCB), were used to analyse their differences in aroma by gas chromatography-olfactometry/mass spectrometry (GC-O/MS). NCB contained more complex volatiles and exhibited a richer aromatic profile compared with BB.
Eduardo Coelho et al.
Food research international (Ottawa, Ont.), 116, 249-257 (2019-02-06)
Cooperage wood is a porous material and beverages exchange compounds with it by penetrating into its pores. This work demonstrates the enrichment of wood with wine during ageing. Three oak varieties were cut into different sized chips and immersed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service