Skip to Content
Merck
All Photos(3)

Key Documents

230057

Sigma-Aldrich

Potassium antimony(III) tartrate hydrate

99.95% trace metals basis

Synonym(s):

Antimony potassium tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4K2O12Sb2 · xH2O
CAS Number:
Molecular Weight:
613.83 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.95% trace metals basis

form

powder

optical activity

[α]20/D +141°, c = 2 in H2O

impurities

≤550.0 ppm Trace Metal Analysis

density

2.6 g/mL at 25 °C (lit.)

SMILES string

O.[K+].[K+].O=C1O[Sb-]23OC1C4O[Sb-]5(OC(C(O2)C(=O)O3)C(=O)O5)OC4=O

InChI

1S/2C4H4O6.2K.H2O.2Sb/c2*5-1(3(7)8)2(6)4(9)10;;;;;/h2*1-2H,(H,7,8)(H,9,10);;;1H2;;/q2*-2;2*+1;;2*+3/p-4

InChI key

BBUFTTIHMIIQLI-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Inocêncio da Luz et al.
Antimicrobial agents and chemotherapy, 53(12), 5197-5203 (2009-09-16)
Diagnostic material from patients with leishmaniasis is generally available as promastigotes, and proper testing for susceptibility to first-line drugs by the intracellular amastigote assay is frequently hampered by the poor infectivity of the promastigotes for the macrophage host cell. Several
T M-H Nguyen et al.
International journal of pharmaceutics, 482(1-2), 21-26 (2014-12-03)
Hyperphosphatemia is one of the main risk factors contributing to morbidity and mortality in patients with end stage renal disease. The demand for a new phosphate binder is continuously increasing since the number of patients suffering under hyperphosphatemia is growing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service