Merck
  • Novel oral phosphate binder with nanocrystalline maghemite-phosphate binding capacity and pH effect.
Novel oral phosphate binder with nanocrystalline maghemite-phosphate binding capacity and pH effect.
International journal of pharmaceutics (2014-12-03)
T M-H Nguyen, R H Müller, M Taupitz, J Schnorr, B Hamm, S Wagner
ABSTRACT
Hyperphosphatemia is one of the main risk factors contributing to morbidity and mortality in patients with end stage renal disease. The demand for a new phosphate binder is continuously increasing since the number of patients suffering under hyperphosphatemia is growing. However, side effects and high pill burden of currently available phosphate binders are the main reasons for low compliance and uncontrolled serum phosphate levels. Therefore, the aim of this study was to develop a novel phosphate binder with a high phosphate binding capacity over the entire gastrointestinal (GI) pH range. This novel phosphate binder C-PAM-10 is based on d-mannose coated nanocrystalline maghemite and belongs to the new class of phosphate binders, called the "iron based agents". It was possible to obtain a phosphate binding product that showed very high phosphate binding capacities with the characteristic of being pH independent at relevant pH ranges. The simulation of a GI passage ranging from pH 1.2 to pH 7.5 showed a 2.5 times higher phosphate binding capacity compared to the commonly used phosphate binder sevelamer carbonate. The simulation of a pH sensitive coating that releases the iron based phosphate binder at pH values ≥4.5 still showed a very high phosphate binding capacity combined with very low iron release which might decrease iron related side effects in vivo. Therefore, C-PAM-10 and its variations may be very promising candidates as a superior phosphate binder.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Potassium antimony(III) tartrate hydrate, ≥99%
Sigma-Aldrich
Potassium antimony(III) tartrate hydrate, 99.95% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture