Skip to Content
Merck
tert-Butyldimethylsilyl trifluoromethanesulfonate reagent grade, 98%

tert-Butyldimethylsilyl trifluoromethanesulfonate

tert-Butyldimethylsilyl trifluoromethanesulfonate reagent grade, 98%
Synonym(s):
TBDMS triflate, Trifluoromethanesulfonic acid tert-butyldimethylsilylester, tert-Butyldimethylsilyl trifluoromethanesulfonate
Linear Formula:
CF3SO3Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
264.34

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group