Skip to Content
Merck
All Photos(1)

Key Documents

225711

Sigma-Aldrich

Nitric acid

70%, purified by redistillation, ≥99.999% trace metals basis

Synonym(s):

Aqua fortis, Azotic acid, Hydrogen nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352301
eCl@ss:
38060304
PubChem Substance ID:
NACRES:
NA.22

vapor density

1 (vs air)

Quality Level

vapor pressure

8 mmHg ( 20 °C)

Assay

≥99.999% trace metals basis
70%

form

liquid

purified by

redistillation

color

colorless

pH

<1.0

bp

120.5 °C (lit.)

density

1.413 g/mL at 20 °C (lit.)

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitric acid is generally used as a primary reagent for the nitration of organic molecules. It is also used as a strong oxidizing agent in organic synthesis to perform the oxidative transformations.

Application

Nitric acid used as a reagent in:
  • The nitration of aromatic compounds, aromatic heterocycles, alkenes, active methylene carbons, etc.
  • Selective oxidation of organosulfur compounds to corresponding sulfoxides using a solid acid catalyst.
  • The oxidation of alcohols, aldehydes, esters, and heteroatoms.
  • The aromatization and dehydrogenation reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chemoselective sulfoxidation by H2O2 or HNO3 using a phosphate impregnated titania catalyst
Bharadwaj SK, et al.
Tetrahedron Letters, 50(27), 3767-3771 (2009)
Quantification of Sodium Accumulation in Arabidopsis thaliana Using Inductively Coupled Plasma Optical Emission Spectrometery (ICP-OES)
Choi WG and Gilroy S
Bio-protocol, 5(16) (2015)
Trace metal contents of three tricalcium silicate materials: MTA Angelus, Micro Mega MTA and Bioaggregate
Kum KY,et al.
International Endodontic Journal, 47(7), 704-710 (2014)
Nitric acid in the presence of P2O5 supported on silica gel?a useful reagent for nitration of aromatic compounds under solvent-free conditions
Hajipour AR and Ruoho AE
Tetrahedron Letters, 46(48), 8307-8310 (2005)
Controlled nitration of anisole over HNO3/PO4/MoO3/SiO2/solvent systems
Adamiak J
J. Mol. Catal. A: Chem., 407, 81-86 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service