Skip to Content
Merck
Nitric acid 65%, univAR™

Nitric acid

Nitric acid 65%, univAR™
Synonym(s):
Nitric acid solution
Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Package Size
Special Grade