Skip to Content
Merck
All Photos(1)

Key Documents

219819

Sigma-Aldrich

2-Hydroxyisocaproic acid

99%

Synonym(s):

2-Hydroxy-4-methylvaleric acid, DL-leucic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

78-80 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

CC(C)CC(O)C(O)=O

InChI

1S/C6H12O3/c1-4(2)3-5(7)6(8)9/h4-5,7H,3H2,1-2H3,(H,8,9)

InChI key

LVRFTAZAXQPQHI-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction-spectrophotometric determination of antimony (V) with 2-hydroxyisocaproic acid and citrate, with application to differential determination of antimony (V) and antimony (III).
SATO S and UCHIKAWA S.
Analytical Sciences, 2, 47-47 (1986)
Polymer, Assisted Solution Phase Synthesis of tr, Ketoamides.
Parlow JJ, et al.
High-Throughput Synthesis: Principles and Practices, 143-143 (2001)
K D Webster et al.
Archives of biochemistry and biophysics, 273(2), 562-571 (1989-09-01)
L-Thiomorpholine-3-carboxylic acid (L-TMC) is a cyclized analog of S-(2-chloroethyl)-L-cysteine, which is cytotoxic in vitro and nephrotoxic in vivo. To determine whether L-TMC may play a role in S-(2-chloroethyl)-L-cysteine-induced toxicity, the cytotoxicity of L-TMC was studied in isolated rat kidney cells.
2-Hydroxyisocaproic acid (HICA): a new potential topical antibacterial agent.
M Sakko et al.
International journal of antimicrobial agents, 39(6), 539-540 (2012-04-10)
E Haan et al.
European journal of pediatrics, 144(1), 63-65 (1985-05-01)
An 8-year-old boy with a short gut had six episodes of metabolic acidosis and neurological dysfunction over a 1 month period. The neurological features consisted of a depressed conscious state, confusion, aggressive behaviour, slurred speech and ataxia. The organic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service