Skip to Content
Merck
All Photos(1)

Key Documents

218847

Sigma-Aldrich

Boron trifluoride methyl etherate

Synonym(s):

(Dimethyl ether)trifluoroboron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · O(CH3)2
CAS Number:
Molecular Weight:
113.87
Beilstein:
3613739
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

autoignition temp.

455 °F

expl. lim.

21.6 %

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

refractive index

n20/D 1.308 (lit.)

bp

126-127 °C (lit.)

mp

−15 °C (lit.)

density

1.239 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

C[O+](C)[B-](F)(F)F

InChI

1S/C2H6BF3O/c1-7(2)3(4,5)6/h1-2H3

InChI key

FOWZWMGLMYVYDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Deconvoluting the chemical reaction kinetics of platinum-silver ligand exchange
  • Synthesis of neurogenic ganglioside LLG-3 from starfish via glycosylation reactions
  • Synthesis of peloruside via lactonization and relay ring-closing metathesis cyclization
  • FRET for monitoring Pd-catalyzed nucleophilic allylic substitution reactions
  • Single molecuel imaging of fluorophore-tagged palldium(II) complexes
  • β-elimination and rearrangement reactions for synthesis of exocyclic vinyl ethers

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Kidney, Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service