Skip to Content
Merck
All Photos(1)

Key Documents

218235

Sigma-Aldrich

(−)-Menthone

90%

Synonym(s):

(1R,4S)-p-Menthan-3-one, (2S,5R)-2-Isopropyl-5-methylcyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2041368
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.5 mmHg ( 20 °C)

Assay

90%

form

liquid

optical activity

[α]20/D −20°, neat

impurities

5% isomenthone

refractive index

n20/D 1.45 (lit.)

bp

207-210 °C (lit.)

density

0.893 g/mL at 20 °C (lit.)

functional group

ketone

SMILES string

CC(C)[C@@H]1CC[C@@H](C)CC1=O

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9+/m1/s1

InChI key

NFLGAXVYCFJBMK-BDAKNGLRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-Menthone is a monoterpenoid compound found in the essential oil extracted from the maturing peppermint (Mentha piperita L.) leaves.

(-)-Menthone can undergo hydrogenation to yield a mixture of (-)-menthol and (+)-neomenthol.

Application

(-)-Menthone may be used to synthesize chiral neomenthyl derivatives, optically pure α-ferrocenylalkylamines and optically active polyalkylcyclohexanones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral perrocenylalkylamines from (-)-menthone.
Siglmuller F, et al.
Tetrahedron, 42(21), 5931-5940 (1986)
Metabolism of Monoterpenes Demonstration of (+)-Neomenthyl-?-d-Glucoside as a Major Metabolite of (-)-Menthone in Peppermint (Mentha Piperita).
Croteau R and Martinkus C.
Plant Physiology, 64(2), 169-175 (1979)
Cerium (III) chloride promoted addition of organometallic reagents to (-)-menthone-preparation of chiral neomenthyl derivatives.
Panev S and Dimitrov V.
Tetrahedron Asymmetry, 11(7), 1517-1526 (2000)
Terpenoids. XLIX. 1 Preparation of Optically Active Polyalkylcyclohexanones from (+)-Pulegone,(-)-Menthone and (-)-Carvone2.
Djerassi C, et al.
Journal of the American Chemical Society, 83(21), 4433-4439 (1961)
Synthesis of liquid menthol by hydrogenation of dementholized peppermint oil over Ni catalysts
Manuale DL, et al.
Quimica nova, 33, 1231-1234 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service