Skip to Content
Merck
All Photos(2)

Key Documents

211680

Sigma-Aldrich

Nigrosin, alcohol soluble

Synonym(s):

Solvent Black 5

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
50415
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47
Pricing and availability is not currently available.

solubility

alcohol: soluble

λmax

565 nm

SMILES string

[N+](=O)([O-])O

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

General description

Nigrosin is a black colored powder.[1] It is widely used to manufacture ink and shoe polish. It is also employed for dyeing leather, wood and textiles. In addition, alcohol soluble nigrosin dye is used in carbon papers, typewriter and ribbons.[2] It is used as an ingredient in rubber repairing lacquer.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Design and validation of a diffuse reflectance and spectroscopic microendoscope with poly(dimethylsioxane)-based phantoms.
Greening GJ
Proc. SPIE: Int. Soc. Opt. Eng. (2015)
Henry Goldschmiedt
Practical Formulas for Hobby Or Profit (1973)
L. Kanerva, P. Elsner, J.E. Wahlberg, H.I. Maibach
Handbook of Occupational Dermatology (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service