Recommended Products
Assay
98%
mp
205-207 °C (lit.)
functional group
carboxylic acid
nitro
SMILES string
Cc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
InChI
1S/C8H6N2O6/c1-4-6(8(11)12)2-5(9(13)14)3-7(4)10(15)16/h2-3H,1H3,(H,11,12)
InChI key
CDVNZMKTJIBBBV-UHFFFAOYSA-N
General description
Synthesis of a series of heterocyclic derivatives of 3,5-dinitro-o-toluic acid has been reported.
Application
3,5-Dinitro-o-toluic acid was used in the synthesis of isoquinolone.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pyridine annelation of o-methyl arylcarboxylic acids with vilsmeier reagents.
Tetrahedron Letters, 24(42), 4607-4610 (1983)
Synthesis of some heterocyclic derivatives of 3, 5-dinitro o-toluic acid.
Journal of Heterocyclic Chemistry, 9, 31-31 (1972)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service