Skip to Content
Merck
All Photos(3)

Key Documents

209465

Sigma-Aldrich

4-Hexylresorcinol

98%

Synonym(s):

Antascarin, Ascarinol, 4-Hexyl-1,3-dihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
194.27
Beilstein:
2048312
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

bp

333-335 °C (lit.)

mp

65-67 °C (lit.)

solubility

water: soluble 2000 part(lit.)
acetone: soluble(lit.)
alcohol: soluble(lit.)
chloroform: soluble(lit.)
diethyl ether: soluble(lit.)
petroleum ether: slightly soluble(lit.)
vegetable oils: soluble(lit.)

SMILES string

CCCCCCc1ccc(O)cc1O

InChI

1S/C12H18O2/c1-2-3-4-5-6-10-7-8-11(13)9-12(10)14/h7-9,13-14H,2-6H2,1H3

InChI key

WFJIVOKAWHGMBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Hexylresorcinol is used as the starting material to synthesize a potent immune suppressor, celastramycin A. It is a precursor to prepare resorcinol-sn-glycerol derivatives, that exhibit high affinity for cannabinoid type 1 receptor. It can also be incorporated as a linker while building catenanes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

388.4 °F

Flash Point(C)

198 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Revised structure and synthesis of celastramycin a, a potent innate immune suppressor.
Kikuchi H, et al.
Organic Letters, 11(8), 1693-1695 (2009)
Seong-Gon Kim et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 69(11), e354-e363 (2011-08-09)
Aerosol deposition is a newly developed technique, and it can deliver the drug from a hydroxyapatite (HA)-coated surface. 4-Hexylresorcinol (4-HR) is a well-known antiseptic. The influence of the 4-HR component of HA coatings on titanium surfaces was studied in vitro
E Arias et al.
Journal of food science, 72(9), C464-C470 (2007-11-24)
We have investigated the mechanism of action of 4-hexylresorcinol (4-HR) and ascorbic acid (AA) on the polyphenol oxidase (PPO) catalyzed oxidation of phenolic substrates. Incubation of PPO with 4-HR diminishes strongly PPO activity. This effect can be erroneously interpreted, due
Seong-Gon Kim et al.
Oncology reports, 29(5), 1835-1840 (2013-02-22)
The bacterial dormancy-inducing factor 4-hexylresorcinol (4-HR) has been shown to have synergistic antitumor effects when used in combination with cisplatin. In the present study, 4-HR was used as a single agent in the squamous carcinoma cell line SCC-9. The results
Resorcinol-sn-glycerol derivatives: novel 2-arachidonoylglycerol mimetics endowed with high affinity and selectivity for cannabinoid type 1 receptor.
Brizzi A, et al.
Journal of Medicinal Chemistry, 54(24), 8278-8288 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service