Skip to Content
Merck
All Photos(2)

Key Documents

177741

Sigma-Aldrich

Perfluorodecanoic acid

98%

Synonym(s):

Nonadecafluorodecanoic acid, Nonadecafluorocapric acid, Perfluorocapric acid, Perfluorodecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)8CO2H
CAS Number:
Molecular Weight:
514.08
Beilstein:
1810811
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

~10 mmHg ( 0 °C)

Assay

98%

bp

218 °C/740 mmHg (lit.)

mp

77-81 °C (lit.)

solubility

methanol: soluble 10%, clear to very slightly hazy, colorless to faintly yellow

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C10HF19O2/c11-2(12,1(30)31)3(13,14)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)29/h(H,30,31)

InChI key

PCIUEQPBYFRTEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The electrochemical mineralization of environmentally persistent perfluorodecanoic acid was studied.

Application

Perfluorodecanoic acid was used to evaluate the effect of nonadecafluorocapric acid on the levels of six thyroid function variables (thyroid stimulating hormone, free and total thyroxine, free and total triiodothyronine and thyroglobulin).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuela Corsini et al.
Toxicology and applied pharmacology, 258(2), 248-255 (2011-11-29)
We have previously shown that PFOA and PFOS directly suppress cytokine secretion in immune cells, with different mechanisms of action. In particular, we have demonstrated a role for PPAR-α in PFOA-induced immunotoxicity, and that PFOS has an inhibitory effect on
Hui Lin et al.
Environmental science & technology, 47(22), 13039-13046 (2013-10-30)
The electrochemical mineralization of environmentally persistent long-chain perfluorinated carboxylic acids (PFCAs), i.e., perfluorononanoic acid (C8F17COOH, PFNA) and perfluorodecanoic acid (C9F19COOH, PFDA) was investigated in aqueous solutions (0.25 mmol L(-1)) over Ti/SnO2-Sb-Ce (SnO2), Ti/SnO2-Sb/Ce-PbO2 (PbO2), and Ti/BDD (BDD) anodes under galvanostatic
Changhui Liu et al.
Environmental science & technology, 45(22), 9758-9764 (2011-10-13)
The effects of exposure concentration on the bioaccumulation of four perfluorinated chemicals (PFCs): perfluorooctanesulfonate (PFOS), perfluoroocanoic acid (PFOA), perfluorononanoic acid (PFNA), and perfluorodecanoic acid (PFDA), was investigated using green mussels, Perna viridis. Mussels were exposed to concentrations of 1 μgL(-1)
Sri Chandana Panchangam et al.
Chemosphere, 77(2), 242-248 (2009-08-12)
Decomposition of perfluorocarboxylic acids (PFCAs) is of prime importance since they are recognized as persistent organic pollutants and are widespread in the environment. PFCAs with longer carbon chain length are particularly of interest because of their noted recalcitrance, toxicity, and
Helena Nilsson et al.
Environmental science & technology, 44(6), 2150-2155 (2010-02-18)
A time trend study focusing on ski waxing technicians' exposure to perfluorinated chemicals (PFCs) from fluorinated wax fumes was performed in 2007/2008. Levels of eight perfluorocarboxylates and three perfluorosulfonates were analyzed in monthly blood samples from eight technicians. Samples were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service