Skip to Content
Merck
All Photos(1)

Key Documents

206288

Sigma-Aldrich

Sodium hexachlororhodate(III)

Synonym(s):

Rhodium sodium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3RhCl6
CAS Number:
Molecular Weight:
384.59
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

Quality Level

SMILES string

[Na+].[Na+].[Na+].Cl[Rh-3](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.3Na.Rh/h6*1H;;;;/q;;;;;;3*+1;+3/p-6

InChI key

YDEXHLGYVJSKTN-UHFFFAOYSA-H

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faqrul A Chowdhury et al.
Nature communications, 9(1), 1707-1707 (2018-04-29)
The conversion of solar energy into chemical fuels can potentially address many of the energy and environment related challenges we face today. In this study, we have demonstrated a photochemical diode artificial photosynthesis system that can enable efficient, unassisted overall
Zhirong Zhang et al.
Nature communications, 11(1), 1215-1215 (2020-03-07)
Single-atom catalysts (SACs) exhibit intriguing catalytic performance owing to their maximized atom utilizations and unique electronic structures. However, the reported strategies for synthesizing SACs generally have special requirements for either the anchored metals or the supports. Herein, we report a
Xinyan Dai et al.
Frontiers in chemistry, 6, 494-494 (2018-11-06)
Silica nanoparticles, optically transparent in the visible spectral region, represent a class of dielectric antenna to tune the propagation and local field distribution of the visible light through surface scattering while the energy loss is minimized. The light scattering on
Trevor B Demille et al.
Frontiers in chemistry, 6, 411-411 (2018-09-27)
Micro- and nanoscale ZnO tetrapods provide an attractive support for metallic nanostructures since they can be inexpensively produced using the flame transport method and nanoparticle synthesis schemes can take advantage of a coupled response facilitated by the formation of a
Kai Liu et al.
Chemistry, an Asian journal, 15(20), 3356-3364 (2020-08-25)
Defective metal nanostructures have attracted great attention due to the striking catalytic behavior of the defect sites. Atypical metal nanocrystals generated from attached nuclei can accommodate abundant grain boundaries (GBs) and twin boundaries (TBs). However, the understanding of their growth-mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service