Skip to Content
Merck
All Photos(1)

Key Documents

205214

Sigma-Aldrich

4-Bromobenzylamine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2NH2 · HCl
CAS Number:
Molecular Weight:
222.51
Beilstein:
3693458
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

274-276 °C (lit.)

functional group

amine

SMILES string

Cl[H].NCc1ccc(Br)cc1

InChI

1S/C7H8BrN.ClH/c8-7-3-1-6(5-9)2-4-7;/h1-4H,5,9H2;1H

InChI key

BVYUYDBWQMSOKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromobenzylamine hydrochloride was used to study the characteristics of C-terminal polyamine modification in the presence of protease and amine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Toshiyuki Ito et al.
Biological & pharmaceutical bulletin, 30(10), 1838-1843 (2007-10-06)
We recently reported that C-terminal polyamine modification occurs when proteins are digested with trypsin in the presence of polyamine [Biochem. Biophys. Res. Commun., 356, 159-162 (2007)]. In the present study, the characteristics of this C-terminal modification in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service