197653
2-(3,4-Dimethoxyphenyl)ethanol
98%
Synonym(s):
3,4-Dimethoxybenzeneethanol, 3,4-Dimethoxyphenethyl alcohol, Homoveratryl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
(CH3O)2C6H3CH2CH2OH
CAS Number:
Molecular Weight:
182.22
Beilstein:
909742
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
bp
172-174 °C/17 mmHg (lit.)
mp
46-49 °C (lit.)
functional group
hydroxyl
SMILES string
COc1ccc(CCO)cc1OC
InChI
1S/C10H14O3/c1-12-9-4-3-8(5-6-11)7-10(9)13-2/h3-4,7,11H,5-6H2,1-2H3
InChI key
SRQAJMUHZROVHW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
2-(3,4-Dimethoxyphenyl)ethanol (Homoveratryl alcohol) was used as a model compound in the delignification mechanism studies of Mn(III)-substituted polyoxometalates (MSP).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Oxygen bleaching of kraft pulp catalysed by Mn (III)-substituted polyoxometalates.
Gaspar A, et al.
Applied Catalysis A: General, 239(1), 157-168 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service