Skip to Content
Merck
All Photos(1)

Key Documents

196681

Sigma-Aldrich

4-(Trifluoromethyl)benzonitrile

99%

Synonym(s):

α,α,α-Trifluoro-p-tolunitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CN
CAS Number:
Molecular Weight:
171.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

refractive index

n20/D 1.4583 (lit.)

bp

80-81 °C/20 mmHg (lit.)

mp

39-41 °C (lit.)

density

1.278 g/mL at 25 °C (lit.)

functional group

fluoro
nitrile

SMILES string

FC(F)(F)c1ccc(cc1)C#N

InChI

1S/C8H4F3N/c9-8(10,11)7-3-1-6(5-12)2-4-7/h1-4H

InChI key

DRNJIKRLQJRKMM-UHFFFAOYSA-N

General description

4-(Trifluoromethyl)benzonitrile is a key intermediate in the synthesis of fluvoxamine. It participates in nickel-catalyzed arylcyanation reaction of 4-octyne.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Hirata et al.
Journal of the American Chemical Society, 131(31), 10964-10973 (2009-09-03)
Allyl cyanides are found to add across alkynes in the presence of a nickel/P(4-CF(3)-C(6)H(4))(3) catalyst to give polysubstituted 2,5-hexadienenitriles with defined stereo- and regiochemistry. Use of AlMe(2)Cl or AlMe(3) as a Lewis acid cocatalyst accelerates the reaction and expands the
Improving palladium-catalyzed cyanation of aryl halides: development of a state-of-the-art methodology using potassium hexacyanoferrate (II) as cyanating agent.
Schareina T, et al.
Journal of Organometallic Chemistry, 689(24), 4576-4583 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service