Skip to Content
Merck
All Photos(1)

Key Documents

193801

Sigma-Aldrich

4,4′-Di-tert-butylbiphenyl

99%

Synonym(s):

1-tert-Butyl-4-(4-tert-butylphenyl)benzene, 4,4′-Bis(1,1-dimethylethyl)-1,1′-biphenyl, 4,4′-Di-tert-butyl-1,1′-biphenyl, DBB, p,p′-Di-tert-butylbiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4C6H4C(CH3)3
CAS Number:
Molecular Weight:
266.42
Beilstein:
2095855
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

190-192 °C/13 mmHg (lit.)

mp

126-130 °C (lit.)

SMILES string

CC(C)(C)c1ccc(cc1)-c2ccc(cc2)C(C)(C)C

InChI

1S/C20H26/c1-19(2,3)17-11-7-15(8-12-17)16-9-13-18(14-10-16)20(4,5)6/h7-14H,1-6H3

InChI key

CDKCEZNPAYWORX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-Di-tert-butylbiphenyl along with lithium catalyzes:
  • reaction of chloromethyl ethyl ether and different carbonyl compounds to yield corresponding hydroxyethers
  • reductive opening of N-phenylazetidine

Application

4,4′-Di-tert-butylbiphenyl was used in production of homoallylic amine derivatives. It was also used in the preparation of lithium di-tert-butylbiphenylide, a radical anion, superior to sodium or lithium naphthalenides for metalation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 55, 1528-1528 (1990)
4, 4′-Di-tert-butylbiphenyl-catalysed lithiation of chloromethyl ethyl ether: A barbier-type new and easy alternative to ethyl lithiomethyl ether.
Guijarro A and Yus M.
Tetrahedron Letters, 34(21), 3487-3490 (1993)
C E Neipp et al.
The Journal of organic chemistry, 66(2), 531-537 (2001-06-30)
Lithiation of the N-2,4,6-triisopropylbenzenesulfonyl-2-pyrroline (16) and treatment of the resulting cyclic vinyllithium reagent with R2CuCNLi2 produced an acyclic vinyl organometallic species that, when treated with an electrophile (H2O or RX), gave the homoallylic sulfonamides 18a-k in 37-93% yields and in
4, 4'-Di -tert -butylbiphenyl-catalysed reductive opening of azetidines with lithium: A direct preparation of 3, N-dilithioalkylamines.
Almena J, et al.
Tetrahedron, 50(19), 5775-5782 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service