Skip to Content
Merck
All Photos(1)

Key Documents

193283

Sigma-Aldrich

2-Amino-4-tert-butylphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(NH2)OH
CAS Number:
Molecular Weight:
165.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

160-163 °C (lit.)

SMILES string

CC(C)(C)c1ccc(O)c(N)c1

InChI

1S/C10H15NO/c1-10(2,3)7-4-5-9(12)8(11)6-7/h4-6,12H,11H2,1-3H3

InChI key

RPJUVNYXHUCRMG-UHFFFAOYSA-N

Application

2-Amino-4-tert-butylphenol can be used as a reactant to prepare:
  • 4-tert-butyl-2-[(pyridylmethylene)amino]phenol intermediates, which are used to synthesize biologically important 2-(pyridyl)benzoxazole derivatives.
  • Prolinamide phenols, as efficient hydrophobic organocatalysts for direct asymmetric aldol reaction aldehydes and ketones in water.
  • N-(2-hydroxy-4-tert-butylphenyl)-acetamide, a key intermediate to prepare uranylsalophene derivatives which can be used as selective receptors in anion sensitive membrane sensors.
  • Poly(2-amino-4-tert-butylphenol) [poly(2A-4TBP)] by electrochemical or chemical oxidative polymerization reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rationally designed 4-phenoxy substituted prolinamide phenols organocatalyst for the direct aldol reaction in water.
Zhang S-P, et al.
Tetrahedron Letters, 50(11), 1173-1176 (2009)
Neutral anion receptors: synthesis and evaluation as sensing molecules in chemically modified field effect transistors.
Antonisse MMG, et al.
The Journal of Organic Chemistry, 62(26), 9034-9038 (1997)
Rationally designed 4-phenoxy substituted prolinamide phenols organocatalyst for the direct aldol reaction in water
Z Shu-peng, et al.
Tetrahedron Letters, 50(11), 1173-1176 (2009)
Synthesis of 2-(2-, 3-, and 4-pyridyl) benzoxazoles by the reaction of phenolic schiff bases with thianthrene cation radical
Park Myeong Soon, et al.
Journal of Heterocyclic Chemistry, 39(6), 1279-1281 (2002)
Synthesis of 2-(2-, 3-, and 4-pyridyl) benzoxazoles by the reaction of phenolic Schiff bases with thianthrene cation radical.
Park MS, et al.
Journal of Heterocyclic Chemistry, 39(6), 1279-1282 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service