Skip to Content
Merck
All Photos(2)

Key Documents

182214

Sigma-Aldrich

Poly(methyl acrylate) solution

average Mw ~40,000 by GPC

Synonym(s):

Acrylic acid methyl ester polymer, Methyl acrylate homopolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H6O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

form

liquid

autoignition temp.

~896 °F

mol wt

average Mw ~40,000 by GPC

concentration

~40 wt. % (in toluene)

refractive index

n20/D 1.494

transition temp

Tg 9 °C

density

1.22 g/mL at 25 °C

SMILES string

COC(=O)C=C

InChI

1S/C4H6O2/c1-3-4(5)6-2/h3H,1H2,2H3

InChI key

BAPJBEWLBFYGME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

PMA/methylamine borane (MeAB) composites, prepared by solution blending process finds uses as a hydrogen storage material with better dehydrogenation property compared to MeAB.

Other Notes

Unit weight does not include weight of solvent.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zanhua Liu et al.
Inflammation, 39(3), 1090-1098 (2016-04-05)
Epilepsy, which affects about 1 % of the population worldwide, leads to poor prognosis and increased morbidity. However, effective drugs providing satisfactory control on seizure relapse were rare, which encouraged more etiological studies. Whether inflammation is one of key events underlying
Decreasing the thermal dehydrogenation temperature of methylamine borane (MeAB) by mixing with poly (methyl acrylate)(PMA).
Yang Z, et al.
International Journal of Hydrogen Energy, 37(9), 7638-7644 (2012)
A soft hydrogen storage material: poly(methyl acrylate)-confined ammonia borane with controllable dehydrogenation.
Jianzhi Zhao et al.
Advanced materials (Deerfield Beach, Fla.), 22(3), 394-397 (2010-03-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service