Skip to Content
Merck
All Photos(2)

Key Documents

182087

Sigma-Aldrich

Poly(ethyl methacrylate)

average Mw ~515,000 by GPC, powder

Synonym(s):

Ethyl methacrylate homopolymer, Poly(ethyl 2-methyl-2-propeonate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2C2H5)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mw ~515,000 by GPC

refractive index

n20/D 1.485

inherent viscosity

0.80 dL/g(lit.)

transition temp

Tg 63 °C

density

1.11 g/mL at 25 °C

SMILES string

CCOC(=O)C(C)=C

InChI

1S/C6H10O2/c1-4-8-6(7)5(2)3/h2,4H2,1,3H3

InChI key

SUPCQIBBMFXVTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PEMA is known to exhibit biocompatibility and in vivo stability in its various use in implants and medical devices. Sorption of low solubility gases in PEMA was observed to be comparatively lower that polymers with high transition temperature.

Application

Poly(ethyl methacrylate) (PEMA) may be used to prepare polymer blends with polystyrene using organically modified clay as emulsifier.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emulsifying effect of dimethyldioctadecylammonium-hectorite in polystyrene/poly (ethyl methacrylate) blends.
Voulgaris D and Petridis D
Polymer, 43(8), 2213-2218 (2002)
Gas sorption and permeation in poly (ethyl methacrylate).
Chiou J S and Paul D R
Journal of Membrane Science , 45(1-2), 167-189 (1989)
Assessment of the parameters influencing the fiber characteristics of electrospun poly (ethyl methacrylate) membranes.
Gomez-Tejedor J A, et al.
European Polymer Journal, 47(2), 119-129 (2011)
Lidia Yileng Tay et al.
Gerodontology, 29(2), e275-e280 (2011-02-03)
To evaluate the effect of water storage time on the cytotoxicity of soft liners. Sample discs of soft liners Dentusoft, Dentuflex, Trusoft, Ufi-Gel-P and denture base acrylic resin Lucitone-550 were prepared and divided into four groups: GN: No treatment, G24:
Hema Kanathila et al.
Indian journal of dental research : official publication of Indian Society for Dental Research, 22(4), 613-613 (2011-11-30)
The presence of Candida albicans on the fitting surface of the denture is a major causative factor in denture stomatitis. A treatment method is by combining tissue conditioner and antifungal agents. The main objective of this study is to test

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service