181587
Poly(tert-butyl methacrylate)
average Mw ~170,000, crystalline
Synonym(s):
tert -Butyl methacrylate homopolymer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C8H14O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
crystalline
Quality Level
mol wt
average Mw ~170,000
refractive index
n20/D 1.4638 (lit.)
transition temp
Tg 107 °C
solubility
8.65 (solubility parameter)
density
1.022 g/mL at 25 °C (lit.)
SMILES string
CC(=C)C(=O)OC(C)(C)C
InChI
1S/C8H14O2/c1-6(2)7(9)10-8(3,4)5/h1H2,2-5H3
InChI key
SJMYWORNLPSJQO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
These triblock terpolymers precursors may be used for the preparation of novel multi-compartmental core-crosslinked polymeric nanoparticles for various nanostructured systems.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis, characterization, and bulk crosslinking of polybutadiene-block-poly (2-vinyl pyridine)-block-poly (tert-butyl methacrylate) block terpolymers.
Schacher F, et al.
Polymer, 51(9), 2021-2032 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service