Skip to Content
Merck
All Photos(1)

Key Documents

152544

Sigma-Aldrich

4-Vinyl-1-cyclohexene 1,2-epoxide, mixture of isomers

98%

Synonym(s):

1,2-Epoxy-4-vinylcyclohexane, 1-Vinyl-3,4-epoxycyclohexane, 3,4-Epoxycyclohexylethylene, 3-Ethenyl-7-oxabicyclo[4.1.0]heptane, Vinyl-3,4-epoxycyclohexane, Vinylcyclohexene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O
CAS Number:
Molecular Weight:
124.18
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

169 °C (lit.)

density

0.952 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC1CCC2OC2C1

InChI

1S/C8H12O/c1-2-6-3-4-7-8(5-6)9-7/h2,6-8H,1,3-5H2

InChI key

SLJFKNONPLNAPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelli L Hvorecny et al.
Structure (London, England : 1993), 25(5), 697-707 (2017-04-11)
Pseudomonas aeruginosa secretes an epoxide hydrolase with catalytic activity that triggers degradation of the cystic fibrosis transmembrane conductance regulator (CFTR) and perturbs other host defense networks. Targets of this CFTR inhibitory factor (Cif) are largely unknown, but include an epoxy-fatty

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service