180106
3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
mp
50-53 °C (lit.)
functional group
carboxylic acid
SMILES string
OC(=O)C(C(F)(F)F)C(F)(F)F
InChI
1S/C4H2F6O2/c5-3(6,7)1(2(11)12)4(8,9)10/h1H,(H,11,12)
InChI key
RAEAYTICAPHWJW-UHFFFAOYSA-N
General description
The behavior of 3,3,3-trifluoro-2-(trifluoromethyl)propionic acid was studied using ion-exclusion chromatography.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of chromatography. A, 884(1-2), 93-103 (2000-08-05)
Ion-exclusion chromatography has been successfully applied to the separation of a number of polyfluorocarboxylic acids. The separation of various mono- and dibasic polyfluorocarboxylic acids having a different alkyl group was investigated using a polymethacrylate-based weakly acidic cation-exchange resin (TSK gel
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service