Skip to Content
Merck
All Photos(1)

Key Documents

17803

Sigma-Aldrich

3-(4-Hydroxy-3-methoxyphenyl)propionic acid

≥96.0% (T)

Synonym(s):

Hydroferulic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
Beilstein:
2110370
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (T)

form

powder

mp

87-93 °C

functional group

carboxylic acid

SMILES string

COc1cc(CCC(O)=O)ccc1O

InChI

1S/C10H12O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2,4,6,11H,3,5H2,1H3,(H,12,13)

InChI key

BOLQJTPHPSDZHR-UHFFFAOYSA-N

Related Categories

Application

3-(4-Hydroxy-3-methoxyphenyl)propionic acid (hydroferulic acid) was used to inhibit prostaglandin E(2) production.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oscar J Lara-Guzmán et al.
Free radical biology & medicine, 160, 604-617 (2020-08-04)
Oxylipins are considered biomarkers related to cardiovascular diseases (CVDs). They are generated in vivo via the oxygenation of polyunsaturated fatty acids as a result of oxidative stress and inflammation. Oxylipins are involved in vascular functions and are produced during foam
Xuhuiqun Zhang et al.
Molecules (Basel, Switzerland), 25(20) (2020-10-22)
Red raspberries (RRB) are high in anthocyanin- and ellagitannin- type (poly)phenols. This study aimed to investigate the effect of 4-week RRB supplementation on (poly)phenolic metabolism in adults with prediabetes and insulin-resistance (PreDM-IR); and whether adding fructo-oligosaccharides (FOS), prebiotics, would augment
Lee E Hunt et al.
Biomolecules, 11(1) (2021-01-06)
There is ongoing interest in exploiting the antioxidant activity and other medicinal properties of natural monophenolic/polyphenolic compounds, but their generally low aqueous solubility limits their applications. Numerous studies have been undertaken to solubilize such compounds via supramolecular derivatization with co-crystal
Mar Larrosa et al.
Molecular nutrition & food research, 53(8), 1044-1054 (2009-06-27)
The polyphenols in fruits and vegetables may be partly responsible for the health-promoting effects attributed to fruit and vegetable intake. Although their properties have been relatively well studied, the activity of their metabolites, produced after ingestion, has been poorly investigated.
Maite Domínguez-Fernández et al.
Food & function, 12(5), 1996-2011 (2021-02-05)
The aim of this study was to evaluate the bioaccessibility of (poly)phenolic compounds in Tudela artichokes (Cynara scolymus cv. Blanca de Tudela) after an in vitro gastrointestinal digestion and the effect of the human colonic microbiota. A total of 28

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service