Skip to Content
Merck
All Photos(1)

Key Documents

102601

Sigma-Aldrich

3,4-Dihydroxyhydrocinnamic acid

98%

Synonym(s):

3-(3,4-Dihydroxyphenyl)propionic acid, Hydrocaffeic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH2CO2H
CAS Number:
Molecular Weight:
182.17
Beilstein:
2213449
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

mp

136-139 °C (lit.)

SMILES string

OC(=O)CCc1ccc(O)c(O)c1

InChI

1S/C9H10O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1,3,5,10-11H,2,4H2,(H,12,13)

InChI key

DZAUWHJDUNRCTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dihydroxyhydrocinnamic acid (DHCA) belongs to the class of monomers known as hydroxycinnamic acids. DHCA, also known as hydrocaffeic acid, can be polymerized to form various caffeic acid-based polymers. These polymers can be used in the development of coatings, films, and packaging materials due to their antioxidant and antimicrobial properties. caffeic acid-based polymers have also shown potential applications in drug delivery systems, tissue engineering, and encapsulation of bioactive compounds. Additionally, these polymers can be incorporated into cosmetic and personal care products due to their antioxidant and anti-inflammatory properties.

Application

3,4-Dihydroxyhydrocinnamic acid(DHCA) can be used as a monomer to prepare muscle-mimetic, bioadhesive polymers. The strong adhesive action of the polymers is due to interactions of catechol groups present at the end of the polymer terminal chain and substrate surface.
It can also be used for thesurface functionalization of nanoparticles. The functionalization with DHCA notonly improves the water solubility of nanoparticles but also provides a platformfor further modification due to the presence of surface carboxyl groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reina Aoki et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(46), 16603-16610 (2011-11-18)
The ability to detect harmful chemicals rapidly is essential for the survival of all animals. In Caenorhabditis elegans (C. elegans), repellents trigger an avoidance response, causing animals to move away from repellents. Dihydrocaffeic acid (DHCA) is a water-soluble repellent and
Lee E Hunt et al.
Biomolecules, 11(1) (2021-01-06)
There is ongoing interest in exploiting the antioxidant activity and other medicinal properties of natural monophenolic/polyphenolic compounds, but their generally low aqueous solubility limits their applications. Numerous studies have been undertaken to solubilize such compounds via supramolecular derivatization with co-crystal
Kebba Sabally et al.
Journal of biotechnology, 127(1), 167-176 (2006-08-15)
Lipase-catalyzed transesterification reaction of dihydrocaffeic acid (DHCA) with flaxseed oil in organic solvent media was investigated. Using equal molar concentration of DHCA and flaxseed oil, only phenolic monoacylglycerols were obtained with a transesterification yield (TY) of 18.9%. A 1:4 DHCA
María Monagas et al.
The British journal of nutrition, 102(2), 201-206 (2009-07-10)
Oligomers and polymers of flavan-3-ols (proanthocyanidins) are very abundant in the Mediterranean diet, but are poorly absorbed. However, when these polyphenols reach the colon, they are metabolised by the intestinal microbiota into various phenolic acids, including phenylpropionic, phenylacetic and benzoic
Abhishek Jain et al.
Scientific reports, 9(1), 9191-9191 (2019-06-27)
Gut microbiome plays a vital role in human health. Human fecal and urine metabolome could provide a functional readout of gut microbial metabolism as well as its interaction with host and diet. However, this relationship still needs to be fully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service