Skip to Content
Merck
All Photos(2)

Key Documents

176788

Sigma-Aldrich

Sodium deuteroxide solution

40 wt. % in D2O, 99 atom % D

Synonym(s):

Sodium deuteroxide solution, Sodium hydroxide-d

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOD / D2O
CAS Number:
Molecular Weight:
41.00
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Quality Level

form

liquid

concentration

40 wt. % in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.4196

mass shift

M+1

SMILES string

[Na+].[O-][2H]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1/i/hD

InChI key

HEMHJVSKTPXQMS-DYCDLGHISA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium deuteroxide solution is an isotopic analog of sodium hydroxide. It is used as a deuterium source to produce other deuterated compounds.

Application

Sodium deuteroxide solution is used:
  • To prepare deuterated THPM (tetrahydropyrimidine).
  • To study the preparation, stability and biodegradability of surface-active amide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Zhai et al.
Biochemistry, 53(21), 3486-3501 (2014-05-16)
Two mutations of the phosphodianion gripper loop in chicken muscle triosephosphate isomerase (cTIM) were examined: (1) the loop deletion mutant (LDM) formed by removal of residues 170-173 [Pompliano, D. L., et al. (1990) Biochemistry 29, 3186-3194] and (2) the loop
Oliver Wahl et al.
Journal of pharmaceutical and biomedical analysis, 95, 1-10 (2014-03-19)
For the impurity profiling of the mucolytic and anti-inflammatory drug carbocisteine a high performance liquid chromatographic (HPLC) method using corona charged aerosol detection (CAD) was developed and fully validated following the ICH guideline Q2(R1). The response was linear (R²>0.995) over
Dominique E Williams et al.
Chembiochem : a European journal of chemical biology, 16(10), 1474-1482 (2015-05-09)
With the goal of designing a lysosomal phospholipase mimic, we optimized experimental variables to enhance Ce(IV) -assisted hydrolysis of phosphatidylcholine (PC) liposomes. Our best result was obtained with the chelating agent bis-tris propane (BTP). Similar to the hydrolytic enzyme, Ce(IV)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service