Merck
  • Impurity profiling of carbocisteine by HPLC-CAD, qNMR and UV/vis spectroscopy.
Impurity profiling of carbocisteine by HPLC-CAD, qNMR and UV/vis spectroscopy.
Journal of pharmaceutical and biomedical analysis (2014-03-19)
Oliver Wahl, Ulrike Holzgrabe
ABSTRACT
For the impurity profiling of the mucolytic and anti-inflammatory drug carbocisteine a high performance liquid chromatographic (HPLC) method using corona charged aerosol detection (CAD) was developed and fully validated following the ICH guideline Q2(R1). The response was linear (R²>0.995) over a small concentration range (0.05-0.25 or 0.10-0.60% respectively) and a detection limit of at least 0.03% was registered. The separation was achieved on a mixed mode column combining hydrophobic C18 and strong cation exchange retention mechanisms using a mass spectrometer compatible volatile mobile phase consisting of trifluoroacetic acid 10 mM and acetonitrile 12% (V/V). Impurities, not assessable by HPLC-CAD such as the volatile chloroacetic acid and the unstable cysteine, were determined by quantitative NMR (qNMR) with maleic acid as internal standard and UV/vis spectroscopy after reaction with Ellman's reagent, respectively. Six batches of three different manufacturers were tested by means of those methods. The purity varied from below 99.0 to higher than 99.8 per cent. The major impurities of all batches were the starting material cystine and N,S-dicarboxymethylcysteine being a synthesis by-product.
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Chloroacetic acid, 99%
Sigma-Aldrich
Chloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Chloroacetic acid, PESTANAL®, analytical standard
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets