Skip to Content
Merck
All Photos(2)

Key Documents

17353

Sigma-Aldrich

di-Boc-cystamine

≥98.0% (TLC)

Synonym(s):

Di-Boc-cystamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28N2O4S2
CAS Number:
Molecular Weight:
352.51
Beilstein:
4261397
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

120-122 °C

SMILES string

CC(C)(C)OC(=O)NCCSSCCNC(=O)OC(C)(C)C

InChI

1S/C14H28N2O4S2/c1-13(2,3)19-11(17)15-7-9-21-22-10-8-16-12(18)20-14(4,5)6/h7-10H2,1-6H3,(H,15,17)(H,16,18)

InChI key

HBTMWZADMHBLMY-UHFFFAOYSA-N

Other Notes

Building block for the synthesis of thioethyl-modified peptides

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W.J. Moree et al.
Tetrahedron Letters, 49, 1133-1133 (1993)
Carolina González-Blanco et al.
Materials (Basel, Switzerland), 12(11) (2019-05-31)
(1) Background. Titanium is characterized by its biocompatibility and resistance to stress and fatigue. Treatment with argon plasma may favour growth of human osteoblasts with respect to cell adhesion and proliferation. The aim of this study was to analyse the
T M Winger et al.
Bioconjugate chemistry, 6(3), 323-326 (1995-05-01)
The derivatization of poly(p-(chloromethyl)styrene-co-divinylbenzene) (Merrifield resin) with N-(tert-butoxycarbonyl)-2-aminoethanethiol is presented as a convenient route for the generation of thiol terminated peptides using a solid phase methodology. Maximum resin substitution reached 92% (773 mumol/g) after 24 h. However, at 30 min

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service