Skip to Content
Merck
All Photos(2)

Key Documents

167746

Sigma-Aldrich

3-Nitrobenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CN
CAS Number:
Molecular Weight:
148.12
Beilstein:
637674
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

114-117 °C (lit.)

functional group

nitrile
nitro

SMILES string

[O-][N+](=O)c1cccc(c1)C#N

InChI

1S/C7H4N2O2/c8-5-6-2-1-3-7(4-6)9(10)11/h1-4H

InChI key

RUSAWEHOGCWOPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Nitrobenzonitrile was employed as matrix for matrix assisted ionization vacuum method for mass spectrometery.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bijay Banstola et al.
The Analyst, 142(10), 1672-1675 (2017-04-27)
We have developed a new ionization approach for matrix-assisted ionization with high temporal resolution using an electrically actuated pulsed valve. Matrix and analyte samples are deposited on a thin metal foil and placed at the inlet of an ambient ionization
Ellen D Inutan et al.
Molecular & cellular proteomics : MCP, 12(3), 792-796 (2012-12-18)
The introduction of electrospray ionization (ESI) and matrix-assisted laser desorption/ionization (MALDI) for the mass spectrometric analysis of peptides and proteins had a dramatic impact on biological science. We now report that a wide variety of compounds, including peptides, proteins, and
Robert B Cody
Journal of the American Society for Mass Spectrometry, 29(8), 1594-1600 (2018-05-31)
Matrix-assisted ionization in vacuum (MAIV) was used to analyze the polyphenol content of ten different tea infusions. Nine different Camellia sinensis infusions were analyzed including three green teas, two black teas, two oolong teas, jasmine tea, and white tea. An
Sarah Trimpin et al.
Analytical chemistry, 85(4), 2005-2009 (2013-02-05)
Matrix assisted ionization vacuum (MAIV) is a new ionization method that does not require high voltages, a laser beam, or applied heat and depends only the proper matrix, 3-nitrobenzonitrile (3-NBN), and the vacuum of the mass spectrometer to initiate ionization.
Yusuf Sert et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 107, 248-255 (2013-02-26)
In the present study, the experimental and theoretical harmonic and anharmonic vibrational frequencies of 4-chloro-3-nitrobenzonitrile were investigated. The experimental FT-IR (400-4000 cm(-1)) and μ-Raman spectra (100-4000 cm(-1)) of the molecule in the solid phase were recorded. Theoretical vibrational frequencies and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service