Skip to Content
Merck
All Photos(2)

Key Documents

164674

Sigma-Aldrich

Triethyloxonium hexachloroantimonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3OSbCl6
CAS Number:
Molecular Weight:
437.66
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: C-C Bond Formation

storage temp.

−20°C

SMILES string

CC[O+](CC)CC.Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/C6H15O.6ClH.Sb/c1-4-7(5-2)6-3;;;;;;;/h4-6H2,1-3H3;6*1H;/q+1;;;;;;;+5/p-6

InChI key

QHWUSZRAYQUNCY-UHFFFAOYSA-H

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changxin Chen et al.
Scientific reports, 6, 22203-22203 (2016-02-27)
A p-i-n junction diode based on the selectively doped single-walled carbon nanotube (SWCNT) had been investigated, in which two opposite ends of individual SWCNT channel were doped into the p- and n-type SWCNT respectively while the middle segment of SWCNT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service