Skip to Content
Merck
All Photos(3)

Key Documents

157929

Sigma-Aldrich

Methyl (triphenylphosphoranylidene)acetate

98%

Synonym(s):

(Carbomethoxymethylene)triphenylphosphorane, (Methoxycarbonylmethylene)triphenylphosphorane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P=CHCO2CH3
CAS Number:
Molecular Weight:
334.35
Beilstein:
618430
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

168-172 °C (lit.)

functional group

ester
phosphine

SMILES string

COC(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H19O2P/c1-23-21(22)17-24(18-11-5-2-6-12-18,19-13-7-3-8-14-19)20-15-9-4-10-16-20/h2-17H,1H3

InChI key

NTNUDYROPUKXNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Wittig reagent for the two-carbon homologation of aldehydes to α,β-unsaturated esters. Also used in an efficient synthesis of pyrazoles via reaction with methyl diazoacetate in the presence of triethylamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 63, 1074-1074 (2007)
Tetrahedron Letters, 48, 883-883 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service