154377
2-Benzofuranyl methyl ketone
99%
Synonym(s):
2-Acetylbenzofuran
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
solid
bp
110-113 °C/3 mmHg (lit.)
mp
70-72 °C (lit.)
functional group
ketone
SMILES string
CC(=O)c1cc2ccccc2o1
InChI
1S/C10H8O2/c1-7(11)10-6-8-4-2-3-5-9(8)12-10/h2-6H,1H3
InChI key
YUTFQTAITWWGFH-UHFFFAOYSA-N
General description
Microwave assisted Claisen-Schmidt condensation of 2-acetylbenzofuran was studied.
Application
2-Benzofuranyl methyl ketone ( 2-Acetylbenzofuran) has been used to synthesize many benzofuran derivatives. It is also used to prepare fluorinated chalcones.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tejpal Singh Chundawat et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 10(4), 409-417 (2013-08-06)
A series of new analogs fluorine containing heterocyclic system viz. 4-benzofuran-2-yl-6-phenyl-pyrimidin-2- ylamine has been synthesized and evaluated for in vitro antibacterial and antifungal activities. Microwave assisted Claisen-Schmidt condensation of 2-acetylbenzofuran (3) with fluorinated benzaldehydes (4) afforded corresponding fluorinated chalcones (5a-g).
Samia M Rida et al.
Archives of pharmacal research, 29(10), 826-833 (2006-11-24)
Previously, we synthesized and evaluated several benzofuran derivatives containing heterocyclic ring substituents linked to the benzofuran nucleus at C-2 by a two- to four-atom spacer as potential anti-HIV-1, anticancer and antimicrobial agents. Among these derivatives, NSC 725612 and NSC 725716
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service