Skip to Content
Merck
All Photos(1)

Key Documents

15408

Sigma-Aldrich

N-Boc-1,3-propanediamine

≥97.0% (GC/NT)

Synonym(s):

N-Boc-1,3-diaminopropane, tert-Butyl N-(3-aminopropyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONH(CH2)3NH2
CAS Number:
Molecular Weight:
174.24
Beilstein:
3588328
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC/NT)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.454 (lit.)
n20/D 1.459

bp

203 °C (lit.)

mp

22 °C (lit.)

density

0.998 g/mL at 20 °C (lit.)

functional group

Boc
amine

SMILES string

NCCCNC(OC(C)(C)C)=O

InChI

1S/C8H18N2O2/c1-8(2,3)12-7(11)10-6-4-5-9/h4-6,9H2,1-3H3,(H,10,11)

InChI key

POHWAQLZBIMPRN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Tackling vancomycin-resistant bacteria with ′lipophilic–vancomycin–carbohydrate conjugates′: This study discusses the synthesis of derivatives using N-Boc-1,3-propanediamine to develop new antibacterial agents targeting resistant bacterial strains (Yarlagadda et al., 2015).
  • Sulfonamides differing in the alkylamino substituent length–Synthesis, electrochemical characteristic, acid-base profile and complexation properties: The study involves N-Boc-1,3-propanediamine in the synthesis of novel sulfonamide derivatives with potential biochemical applications (Ciesielska et al., 2022).
  • Direct α-alkylation of primary aliphatic amines enabled by CO2 and electrostatics: Research demonstrating selective α-alkylation of N-Boc-1,3-propanediamine, highlighting a novel method in organic synthesis (Ye et al., 2018).

Other Notes

Synthesis of spermidine analogues; Preparation of pharmacologically active compounds.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Huang et al.
Journal of medicinal chemistry, 35(13), 2414-2418 (1992-06-26)
A number of substrate analogues of N8-acetylspermidine (N8-AcSpd) (16) and chemical modifying agents containing metal coordinating ligands were assayed as inhibitors of the cytoplasmic enzyme N8-AcSpd deacetylase from rat liver. The enzyme is inhibited by metal chelators, several omega-amino-substituted carboxylic
Syota Kawaguchi et al.
The Journal of reproduction and development, 59(3), 219-224 (2013-02-07)
Luteoprotective mechanisms of luteinizing hormone (LH) involved in the maintenance of bovine corpus luteum (CL) function have not been completely clarified. Since antioxidant enzymes are well documented as antiapoptotic factors in the CL of many mammals, we hypothesized that the
Syota Kawaguchi et al.
The Journal of reproduction and development, 59(3), 225-230 (2013-01-30)
Luteinizing hormone (LH) regulates several ovarian functions. However, the luteoprotective mechanisms of LH involved in the maintenance of bovine corpus luteum (CL) function are not well understood. Since prostaglandin F2α (PGF), PGE2 and progesterone (P4) are well documented as antiapoptotic
B. Plouvier et al.
Heterocycles, 32, 693-693 (1991)
Hironori Abe et al.
The Journal of reproduction and development, 61(4), 277-286 (2015-05-01)
We recently demonstrated that luteal cells flow out from the ovary via lymphatic vessels during luteolysis. However, the regulatory mechanisms of the outflow of luteal cells are not known. Matrix metalloproteinases (MMPs) can degrade the extracellular matrix and basal membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service