Skip to Content
Merck
All Photos(2)

Key Documents

151831

Sigma-Aldrich

Chloroform-d

99.8 atom % D, contains 1 % (v/v) TMS

Synonym(s):

Deuterochloroform

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
Beilstein:
1697633
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.8 atom % D

Quality Level

Assay

99% (CP)

form

liquid

contains

1 % (v/v) TMS

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.444 (lit.)

bp

60.9 °C (lit.)

mp

−64 °C (lit.)

density

1.500 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]C(Cl)(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform-d (Deuterochloroform, CDCl3) is a deuterated derivative of chloroform. It contains 1%(v/v) TMS (Tetramethylsilane, added as a stabilizer). Quantitative infrared spectral investigations of carbon-deuterium stretching bands of chloroform-d in various organic solvents have been reported. Raman difference spectroscopic studies of mixtures of chloroform-d and liquid chloroform have been conducted to evaluate frequency shifts in the in the ν1 and ν2 bands of CHCl3 and CDCl3.

Application

Chloroform-d has been used as the deuterated solvent for the NMR spectral studies of the corresponding chiral derivatives of β-aminopropanoic acid containing the α-phenylethyl group, which serve as useful starting materials for the asymmetric synthesis of β-amino acids.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of chiral derivatives of ?-Ala containing the a-phenylethyl group: useful starting materials for the asymmetric synthesis of ?-amino acids
Guzman-Mejia R, et al.
Nature Protocols, 2(11), 2759-2766 (2007)
Isotopic dilution studies of the chloroform-chloroform-d system by Raman difference spectroscopy.
Laane J and Kiefer W.
J. Chem. Phys. , 73(10), 4971-4975 (1980)
Quantitative Study of the Bonding of Chloroform-d in Various Solvents by Infrared Spectrometry1.
Lord RC, et al.
Journal of the American Chemical Society, 77(5), 1365-1368 (1955)
Ramón Guzmán-Mejía et al.
Nature protocols, 2(11), 2759-2766 (2007-11-17)
The use of microwave (MW) irradiation for the condensation reaction between acetophenone and alpha-phenylethylamine to prepare (R,R)-bis[alpha-phenylethyl]amine results in significantly reduced reaction times relative to the use of conventional heating. In this protocol, a secondary amine, (R,R)-bis(alpha-phenylethyl)amine is treated with
Chong Li et al.
Nature communications, 5, 5709-5709 (2014-12-17)
Photoswitchable fluorescent diarylethenes are promising in molecular optical memory and photonic devices. However, the performance of current diarylethenes is far from satisfactory because of the scarcity of high-speed switching capability and large fluorescence on-off ratio. Here we report a trident

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service