Skip to Content
Merck
All Photos(1)

Key Documents

151769

Sigma-Aldrich

4-Piperidone monohydrate hydrochloride

98%

Synonym(s):

4,4-Piperidinediol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO · HCl · H2O
CAS Number:
Molecular Weight:
153.61
Beilstein:
3916558
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

94-96 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].O=C1CCNCC1

InChI

1S/C5H11NO2.ClH/c7-5(8)1-3-6-4-2-5;/h6-8H,1-4H2;1H

InChI key

BQOLKFJNJCOALF-UHFFFAOYSA-N

Application

4-Piperidone monohydrate hydrochloride was used in the generation of compound libraries based on sub-reactions of an Ugi multicomponent reaction (MCR).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microchip-based synthesis and total analysis systems (?SYNTAS): chemical microprocessing for generation and analysis of compound libraries.
Journal of the Chemical Society. Perkin Transactions 1, 5, 514-518 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service