Skip to Content
Merck
All Photos(1)

Key Documents

15175

Sigma-Aldrich

Bis[tetrakis(hydroxymethyl)phosphonium] sulfate solution

technical, 70-75% in H2O (T)

Synonym(s):

Tetrakis(hydroxymethyl)phosphonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(HOCH2)4P]2(SO4)
CAS Number:
Molecular Weight:
406.28
Beilstein:
8521357
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

concentration

70-75% in H2O (T)

impurities

formaldehyde

SMILES string

[O-]S([O-])(=O)=O.OC[P+](CO)(CO)CO.OC[P+](CO)(CO)CO

InChI

1S/2C4H12O4P.H2O4S/c2*5-1-9(2-6,3-7)4-8;1-5(2,3)4/h2*5-8H,1-4H2;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

YIEDHPBKGZGLIK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yao Yuan et al.
Polymers, 12(1) (2020-01-08)
Epoxy resin (EP) has widespread applications in thermosetting materials with great versatility and desirable properties such as high electrical resistivity and satisfactory mechanical properties. At present, 9,10-Dihydro-9-oxa-10-phosphaphenanthrene-10-oxide (DOPO) is widely applied to EP matrix for high flame resistance. Nevertheless, EP/DOPO
Christian Bayreder et al.
Medical physics, 33(7), 2506-2518 (2006-08-11)
Magnetic resonance (MR)-based polymer gel dosimetry using normoxic polymer gels, represents a new dosimetric method specially suited for high-resolution three-dimensional dosimetric problems. The aim of this study was to investigate the dose response with regard to stability, accuracy, reproducibility, and
Baris Karakullukcu et al.
Journal of biophotonics, 4(10), 740-751 (2011-08-24)
The objective quantitative monitoring of light, oxygen, and photosensitizer is challenging in clinical photodynamic therapy settings. We have previously developed fluorescence differential path-length spectroscopy (FDPS), a technique that utilizes reflectance spectroscopy to monitor microvascular oxygen saturation, blood volume fraction, and
S G Bown et al.
Canadian journal of gastroenterology = Journal canadien de gastroenterologie, 13(5), 389-392 (1999-06-23)
Most applications of photodynamic therapy (PDT) in gastroenterology to date have used porfimer sodium as the photosensitizing agent. For destroying small lesions in the wall of the gastrointestinal tract in inoperable patients, it has proved to be most effective, but
Jian-dong Wang et al.
Zhonghua wai ke za zhi [Chinese journal of surgery], 49(4), 341-345 (2011-05-27)
To study the pharmacokinetics, distribution and excretion of m-THPC in rat models of liver cancer via orthotropic implantation using Walker-256. After an intravenous injection of m-THPC with 0.3 mg/kg, the concentrations of m-THPC in biological specimens were determined by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service