Skip to Content
Merck
All Photos(2)

Key Documents

151483

Sigma-Aldrich

9-Ethyl-3-carbazolecarboxaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

85-87 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc2n(CC)c3ccccc3c2c1

InChI

1S/C15H13NO/c1-2-16-14-6-4-3-5-12(14)13-9-11(10-17)7-8-15(13)16/h3-10H,2H2,1H3

InChI key

QGJXVBICNCIWEL-UHFFFAOYSA-N

Application

9-Ethyl-3-carbazolecarboxaldehyde was used in the synthesis of poly(vinyl acetal)s (PVAcs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agata Głuszyńska et al.
Molecules (Basel, Switzerland), 24(5) (2019-03-13)
Microwave formylation of carbazole derivatives was investigated and 3-monoaldehydes were obtained in high yield. A potential DNA-binding ligand, 3-[(3-ethyl)-2-vinylbenzothiazolium]-9-N-ethyl carbazole iodide, was synthesized and characterized including spectral properties (UV-Vis absorption and fluorescence spectra). The binding selectivity and affinity of three
Poly (vinyl acetal) s containing electron-donor groups: Synthesis in homogeneous phase and their thermal properties.
Fernandez MD, et al.
Reactive functional Polymers, 68(1), 39-56 (2008)
Ramasamy Raj Kumar et al.
Journal of photochemistry and photobiology. B, Biology, 165, 310-327 (2016-11-14)
Formation of ruthenium(II) complexes of the type [RuH(CO)(PPh

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service