Skip to Content
Merck
All Photos(1)

Key Documents

151068

Sigma-Aldrich

2,5-Dimethylbenzaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3CHO
CAS Number:
Molecular Weight:
134.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.544 (lit.)

bp

104.5-106.5 °C/14 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)c(C=O)c1

InChI

1S/C9H10O/c1-7-3-4-8(2)9(5-7)6-10/h3-6H,1-2H3

InChI key

SMUVABOERCFKRW-UHFFFAOYSA-N

General description

The LD vehicle′s emmision factor for 2,5-dimethylbenzaldehyde was studied.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobumitsu Sakai et al.
The Science of the total environment, 586, 1279-1286 (2017-02-27)
Volatile Organic Compounds (VOCs) in indoor air were investigated at 39 private residences in Selangor State, Malaysia to characterize the indoor air quality and to identify pollution sources. Twenty-two VOCs including isomers (14 aldehydes, 5 aromatic hydrocarbons, acetone, trichloroethylene and
Daniel Grosjean et al.
Research report (Health Effects Institute), (107)(107), 57-78 (2002-04-17)
Carbonyls (aldehydes and ketones) continue to receive scientific and regulatory attention as toxic air contaminants, mutagens, and carcinogens. Vehicle emissions are a major source of carbonyls in outdoor air, but information about the nature and magnitude of carbonyl emissions by
Suk Ling Wee et al.
Phytochemistry, 153, 120-128 (2018-06-16)
The plants of the enigmatic genus Rafflesia are well known for their gigantic flowers and their floral features such as pungent floral scent and vivid dark color, which mimics the food/brood sites of carrion. However, information on the pollination biology
Florentina Villanueva et al.
Environmental technology, 37(8), 983-996 (2015-10-29)
This work presents observations of volatile organic compounds (VOCs), including carbonyls, particulate matter 2.5 (PM2.5) (included in the most recent ambient air quality standards because of its harmful effect on health), PM10 and other important pollutants, CO, SO2, NOx and
Jana Rousová et al.
Journal of chromatography. A, 1544, 49-61 (2018-03-06)
Carboxylic acids and aldehydes are present in ambient air particulate matter (PM) originating from both primary emission and secondary production in air and may, due to their polarity have, an impact on formation of cloud condensation nuclei. Their simultaneous determination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service