Skip to Content
Merck
All Photos(1)

Key Documents

148067

Sigma-Aldrich

Glycidyl 4-methoxyphenyl ether

99%

Synonym(s):

2,3-Epoxypropyl 4-methoxyphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O3
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.25

Assay

99%

form

solid

mp

45-48 °C (lit.)

SMILES string

COc1ccc(OCC2CO2)cc1

InChI

1S/C10H12O3/c1-11-8-2-4-9(5-3-8)12-6-10-7-13-10/h2-5,10H,6-7H2,1H3

InChI key

AVWGFHZLPMLKBL-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya'nan Wang et al.
Polymers, 12(5) (2020-05-30)
Ring-opening copolymerization (ROCOP) is an effective means to prepare functionalized polyester. In this work, a type of selenide-containing polyesters with controllable structure, molecular weight, and molecular weight distribution was successfully prepared by ROCOP of γ-selenobutyrolactone and epoxy compounds. The influence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service