Skip to Content
Merck
All Photos(1)

Key Documents

146161

Sigma-Aldrich

5-Methylsalicylic acid

98%

Synonym(s):

2-Hydroxy-5-methylbenzoic acid, p-Cresotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OH)CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
1909076
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

150-154 °C (lit.)

functional group

carboxylic acid

SMILES string

Cc1ccc(O)c(c1)C(O)=O

InChI

1S/C8H8O3/c1-5-2-3-7(9)6(4-5)8(10)11/h2-4,9H,1H3,(H,10,11)

InChI key

DLGBEGBHXSAQOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methylsalicylic acid exhibits a significant cross-reactivity during fluorescent polarization immunoassay for salicylates in serum.

Application

5-Methylsalicylic acid was used in generation of o-sulfate conjugates in situ and their analysis by ultra-performance liquid chromatography-time-of-flight mass spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Koel et al.
Journal of analytical toxicology, 13(6), 358-360 (1989-11-01)
We studied the recently developed Abbott Laboratories fluorescent polarization immunoassay (FPIA) for salicylates in serum. The cross-reactivity test of the assay was performed with 20 substances that showed a similar chemical structure as salicylic acid. A chemical substitution on the
Jong Dae Jang et al.
Polymers, 13(2) (2021-01-10)
Pluronic amphiphilic block copolymers, well known to have a phase behavior can be controlled by external conditions, have a wide range of potential for applications such as nanotemplates or nanobuilding blocks. However, the phase behaviors of Pluronic block copolymer/additive complexes
Jong-Dae Jang et al.
Micromachines, 9(10) (2018-11-15)
The phase behavior of amphiphilic Pluronic block copolymers in aqueous solution is of importance for a broad spectrum of practical applications but has not been fully exploited yet. Here, the phase behavior of the mixture of the Pluronic P65 and
Shabnam Najafi AsliPashaki et al.
Talanta, 195, 807-814 (2019-01-11)
In this research, a simple, rapid, and efficient air assisted - vesicle based microextraction (AAVME) approach was developed for the extraction of phenolic compounds and their analysis in different Melissa officinalis L. samples. The extraction method is based injection an
John B Tyburski et al.
Radiation research, 170(1), 1-14 (2008-06-28)
Gamma-radiation exposure has both short- and long-term adverse health effects. The threat of modern terrorism places human populations at risk for radiological exposures, yet current medical countermeasures to radiation exposure are limited. Here we describe metabolomics for gamma-radiation biodosimetry in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service