All Photos(1)
About This Item
Linear Formula:
H2C=CHCH2C6H2(OCH3)2OH
CAS Number:
Molecular Weight:
194.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95%
form
liquid
refractive index
n20/D 1.548 (lit.)
bp
168-169 °C/11 mmHg (lit.)
density
1.092 g/mL at 25 °C (lit.)
SMILES string
COc1cc(CC=C)cc(OC)c1O
InChI
1S/C11H14O3/c1-4-5-8-6-9(13-2)11(12)10(7-8)14-3/h4,6-7,12H,1,5H2,2-3H3
InChI key
FWMPKHMKIJDEMJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M Sugumaran et al.
Archives of biochemistry and biophysics, 353(2), 207-212 (1998-06-02)
Tyrosinase, which is known to possess both monophenol monooxygenase activity (EC 1.14.18.1, tyrosine, 3,4-dihydroxyphenylalanine:oxygen oxidoreductase) and o-diphenoloxidase activity (EC 1.10.3.1, o-diphenol:oxygen oxidoreductase), has been shown to exhibit other related activities. Recently, a new reaction, viz., oxidative conversion of 2,6-dimethoxyallyl phenol
Bruna Pasqualotto Costa et al.
Journal of ethnopharmacology, 267, 113645-113645 (2020-12-04)
Plant-derived compounds are a reservoir of natural chemicals and can act as drug precursors or prototypes and pharmacological probes. Methoxyeugenol is a natural compound found in plant extracts, such as nutmeg (Myristica fragrans), and it presents anthelmintic, antimicrobial, anti-inflammatory activities.
K Kónya et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 8(6), 454-459 (2002-02-05)
A series of naturally occurring 8.0.4'-neolignans (1a-d, 1g, 2g, 2h) and their analogues (le-f, lh, 1i, 2a-f, 2i) have been synthesized in racemic form starting from commercially available phenols, such as eugenol, isoeugenol and 4-allyl-2,6-dimethoxyphenol and from aromatic aldehydes, such
Supriya Agnihotri et al.
Natural product research, 26(23), 2266-2269 (2012-01-21)
Hydrodistilled oil obtained from the stem bark of Myrica esculenta Buch. Ham. ex D. Don (yield 0.3%) was analysed by capillary GC and GC-MS. The volatile oil consisted mainly of n-hexadecanol (25.2%), eudesmol acetate (21.9%), palmitic acid (11.6%), cis-β-caryophyllene (8.7%)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service