Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.488 (lit.)
bp
74 °C/15 mmHg (lit.)
mp
3-5 °C (lit.)
density
0.980 g/mL at 20 °C
0.971 g/mL at 25 °C (lit.)
functional group
ketone
storage temp.
2-8°C
SMILES string
CC1=CC(=O)CC1
InChI
1S/C6H8O/c1-5-2-3-6(7)4-5/h4H,2-3H2,1H3
InChI key
CHCCBPDEADMNCI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
3-Methyl-2-cyclopentenone undergoes reduction in presence of triethylammonium formate and palladium catalyst to form saturated ketone. It undergoes B(C6F5)3 catalyzed hydrosilation to yield complex reaction mixtures of 1,2 addition products and oligomers.
Application
3-Methyl-2-cyclopentenone was used in the synthesis of 2-hydroxy-3-methyl-2-cyclopentenone.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
149.0 °F - closed cup
Flash Point(C)
65 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A new synthesis of cyclopentenones: dihydrojasmone.
The Journal of Organic Chemistry, 50(12), 2110-2112 (1985)
Palladium-catalyzed reductions of. alpha.,. beta.-unsaturated carbonyl compounds, conjugated dienes, and acetylenes with trialkylammonium formates.
The Journal of Organic Chemistry, 43(20), 3985-3987 (1978)
B(C6F5)3 catalyzed hydrosilation of enones and silyl enol ethers.
Tetrahedron, 58(41), 8247-8254 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service