Skip to Content
Merck
All Photos(1)

Key Documents

145025

Sigma-Aldrich

5-Amino-6-nitroquinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N3O2
CAS Number:
Molecular Weight:
189.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

272-273 °C (dec.) (lit.)

functional group

nitro

SMILES string

Nc1c(ccc2ncccc12)[N+]([O-])=O

InChI

1S/C9H7N3O2/c10-9-6-2-1-5-11-7(6)3-4-8(9)12(13)14/h1-5H,10H2

InChI key

TYBYHEXFKFLRFT-UHFFFAOYSA-N

General description

Electrochemical reduction of 5-amino-6-nitroquinoline has been studied at carbon paste electrode by differential pulse voltammetry, direct current voltammetry, adsorptive stripping voltammetry and HPLC with electrochemical detection.

Application

5-Amino-6-nitroquinoline was used in preparation of imidazoquinoline derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Weisburger et al.
Environmental health perspectives, 67, 121-127 (1986-08-01)
Because mutagens typified by 2-amino-3-methylimidazo[4,5-f]quinoline (IQ) observed in cooked foods are widely consumed, detailed studies of their biochemical and biological properties including carcinogenicity are most important. IQ induces unscheduled DNA synthesis in liver cells, which when taken together with its
Determination of 5-amino-6-nitroquinoline at a carbon paste electrode.
Nemcova L, et al.
Collection of Czechoslovak Chemical Communications, 74(10), 1477-1488 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service