Skip to Content
Merck
All Photos(1)

Key Documents

140244

Sigma-Aldrich

N,N-Dimethylformamide dineopentyl acetal

99%

Synonym(s):

1,1-Dineopentyloxy-N,N-dimethylmethylamine, 1,1-Dineopentyloxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH[OCH2C(CH3)3]2
CAS Number:
Molecular Weight:
231.37
Beilstein:
741992
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

85-87 °C/10 mmHg (lit.)

density

0.829 g/mL at 25 °C (lit.)

functional group

amine
ether

SMILES string

CN(C)C(OCC(C)(C)C)OCC(C)(C)C

InChI

1S/C13H29NO2/c1-12(2,3)9-15-11(14(7)8)16-10-13(4,5)6/h11H,9-10H2,1-8H3

InChI key

KEXFRBIOHPDZQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethylformamide dineopentyl acetal was used in the esterification of Nα-9-fluorenylmethyloxycarbonylamino acids(Fmoc). It was used in the synthesis of 1,3-dialkyl, benzyl and cyclohexyl barbiturate derivatives. It was used as reagent during the synthesis of L-serine and L-cystine stereospecifically labeled with deuterium at the β-position.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of l-serine and l-cystine stereospecifically labeled with deuterium at the ?-position.
Oba M, et al.
Tetrahedron Asymmetry, 17(12), 1890-1894 (2006)
F Albericio et al.
International journal of peptide and protein research, 23(4), 342-349 (1984-04-01)
The attachment of Fmoc-amino acids onto p- alkoxybenzyl alcohol resins via DCC-DMAP coupling suffers from two different problems: formation of dimers and racemization. The use of N,N-dimethylformamide dineopentyl acetal for the preparation of Fmoc- aminoacyloxybenzyl handles is the basis of
R D Budd
Clinical toxicology, 16(2), 189-199 (1980-04-01)
Procedures are described for the on-column gas chromatographic synthesis of various N-alkyl-N-alkyl barbiturates where groups added to the nitrogens are benzyl, cyclohexyl, or straight chains with 1 to 10 carbons. The 1,3-dialkyl barbiturates having methyl, ethyl, n-propyl, n-butyl, cyclohexyl, isopropyl
F Albericio et al.
International journal of peptide and protein research, 26(1), 92-97 (1985-07-01)
Several Fmoc-amino acids have been esterified by use of N,N-dimethylformamide dineopentyl acetal to 2,4,5-trichlorophenyl 3'-(4''-hydroxymethyl-phenoxy)propionate, and the resultant handle derivatives were purified and then quantitatively coupled onto aminomethyl supports. Compared to literature methodology, the present procedure is preferred because: (i)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service